Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 427861000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1066-45-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Strength0.90 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only