Thermo Scientific Chemicals

N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, 98%, Thermo Scientific Chemicals

Catalog number: 427970250
25 g, Each
Thermo Scientific Chemicals

N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, 98%, Thermo Scientific Chemicals

Catalog number: 427970250
25 g, Each
Quantity
Catalog number: 427970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
152120-54-2
IUPAC Name
tert-butyl N-({[(tert-butoxy)carbonyl]imino}(1H-pyrazol-1-yl)methyl)carbamate
Molecular Formula
C14H22N4O4
InChI Key
QFNFDHNZVTWZED-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
86°C to 90°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text