L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 427995000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namedipotassium (2R,3R)-2,3-dihydroxybutanedioate
Molecular FormulaC4H4K2O6
InChI KeyAVTYONGGKAJVTE-OLXYHTOASA-L
SMILES[K+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Molecular Weight (g/mol)226.27
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<4 % (150°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only