Thermo Scientific Chemicals

tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

Catalog number: 428010025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

Catalog number: 428010025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 428010025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1634-04-4
IUPAC Name
2-methoxy-2-methylpropane
Molecular Formula
C5H12O
InChI Key
BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES
COC(C)(C)C
Density
(20/4°C) 0.740 to 0.742
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
UV
at 210 nm A: =<1.0
UV
at 350 nm A: =<0.01

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text