Thermo Scientific Chemicals

Octylmagnesium chloride, 1.4M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428031000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Octylmagnesium chloride, 1.4M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428031000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 428031000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
38841-98-4
IUPAC Name
chloro(octyl)magnesium
Molecular Formula
C8H17ClMg
InChI Key
DPBWWQDKVQLWFK-UHFFFAOYSA-M
SMILES
CCCCCCCC[Mg]Cl
Appearance (Form)
Solution
Titration Grignard
1.2 to 1.6 M
Appearance (Color)
Grey-green to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text