Thermo Scientific Chemicals

3-(Benzyldimethylammonio)propanesulfonate, 95%, Thermo Scientific Chemicals

Catalog number: 428070250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-(Benzyldimethylammonio)propanesulfonate, 95%, Thermo Scientific Chemicals

Catalog number: 428070250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428070250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81239-45-4
IUPAC Name
3-(benzyldimethylazaniumyl)propane-1-sulfonate
Molecular Formula
C12H19NO3S
InChI Key
MEJASPJNLSQOAG-UHFFFAOYSA-N
SMILES
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
NMR
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text