TABS, 99%
Thermo Scientific Chemicals

TABS, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 428081000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54960-65-5
IUPAC Name4-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}butane-1-sulfonic acid
Molecular FormulaC8H19NO6S
InChI KeyVFTZCDVTMZWNBF-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCCCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=98.5 % (On anhydrous basis)
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only