2,2-Dimethyl-4-pentenoic acid, 97%, Thermo Scientific Chemicals
2,2-Dimethyl-4-pentenoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Dimethyl-4-pentenoic acid, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 428090010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16386-93-9
IUPAC Name2,2-dimethylpent-4-enoic acid
Molecular FormulaC7H12O2
InChI KeyBGUAPYRHJPWVEM-UHFFFAOYSA-N
SMILESCC(C)(CC=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4310 to 1.4340 (20°C, 589 nm)
Titration with NaOH>=96.0 %
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only