Thermo Scientific Chemicals

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%, Thermo Scientific Chemicals

Catalog number: 428110100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%, Thermo Scientific Chemicals

Catalog number: 428110100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428110100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1517-82-4
IUPAC Name
5-methyl-2-(propan-2-yl)cyclohexyl 4-methylbenzene-1-sulfinate
Molecular Formula
C17H26O2S
InChI Key
NQICGNSARVCSGJ-UHFFFAOYNA-N
SMILES
CC(C)C1CCC(C)CC1OS(=O)C1=CC=C(C)C=C1
Melting point
100°C to 107°C
HPLC
>=97.5 %
Specific optical rotation
-190° to -204° (20°C, 589 nm) (c=2,acetone)
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text