Thermo Scientific Chemicals

(±)-alpha-Tocopherol, 95%, synthetic

Catalog number: 428125000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

(±)-alpha-Tocopherol, 95%, synthetic

Catalog number: 428125000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428125000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10191-41-0
IUPAC Name
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Molecular Formula
C29H50O2
InChI Key
GVJHHUAWPYXKBD-IEOSBIPESA-N
SMILES
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
Appearance (Color)
Clear colorless to light brown
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text