Methyl linoleate, 99%, Thermo Scientific Chemicals
Methyl linoleate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl linoleate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 428130250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-63-0
IUPAC Namemethyl (9Z,12Z)-octadeca-9,12-dienoate
Molecular FormulaC19H34O2
InChI KeyWTTJVINHCBCLGX-NQLNTKRDSA-N
SMILESCCCCCC=C/CC=C/CCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only