Thermo Scientific Chemicals

Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428141000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428141000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 428141000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
593-04-4
IUPAC Name
1-butoxybutane; trifluoroborane
Molecular Formula
C8H18BF3O
InChI Key
PVKVBDZRILNPJY-UHFFFAOYSA-N
SMILES
FB(F)F.CCCCOCCCC
Appearance (Color)
Light yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Strength
30.0 to 35.0 % (Boron trifluoride)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text