Methyl p-tolyl sulfoxide, 98%, Thermo Scientific Chemicals
Methyl p-tolyl sulfoxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl p-tolyl sulfoxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 428150050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-methanesulfinyl-4-methylbenzene
Molecular FormulaC8H10OS
InChI KeyFEVALTJSQBFLEU-UHFFFAOYNA-N
SMILESCC1=CC=C(C=C1)S(C)=O
Molecular Weight (g/mol)154.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Low melting powder or crystals
Infrared spectrumConforms
NMRconsistent with structure
Melting point41°C to 46°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only