Aluminum acetate, 90%, basic, Thermo Scientific Chemicals
Aluminum acetate, 90%, basic, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum acetate, 90%, basic, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 428190010
Price (USD)/ Each
168.00
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
168.00
Add to cart
Aluminum acetate, 90%, basic, Thermo Scientific Chemicals
Catalog number428190010
Price (USD)/ Each
168.00
-
Add to cart
Chemical Identifiers
CAS142-03-0
IUPAC Name(acetyloxy)(hydroxy)alumanyl acetate
Molecular FormulaC4H7AlO5
InChI KeyHQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILESCC(=O)O[Al](O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Aluminium (Al)15.0 to 18.3 % (ICP)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only