Thermo Scientific Chemicals

Aluminum acetate, 90%, basic, Thermo Scientific Chemicals

Catalog number: 428192500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Aluminum acetate, 90%, basic, Thermo Scientific Chemicals

Catalog number: 428192500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428192500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-03-0
IUPAC Name
(acetyloxy)(hydroxy)alumanyl acetate
Molecular Formula
C4H7AlO5
InChI Key
HQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILES
CC(=O)O[Al](O)OC(C)=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Aluminium (Al)
15.0 to 18.3 % (ICP)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text