Thermo Scientific Chemicals

2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, 97%, Thermo Scientific Chemicals

Catalog number: 428250010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, 97%, Thermo Scientific Chemicals

Catalog number: 428250010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428250010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
331465-71-5
IUPAC Name
2,8,9-tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecan-5-ium
Molecular Formula
C18H40N4P
InChI Key
WFHPXSHLCFHEIA-UHFFFAOYSA-O
SMILES
CC(C)CN1CC[NH+]2CCN(CC(C)C)P1N(CC(C)C)CC2
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Elemental Analysis
60.60 to 65.65 % C
Elemental Analysis
11.02 to 11.94 % H
Elemental Analysis
15.70 to 17.01 % N

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text