Thermo Scientific Chemicals

Phosphorus pentabromide, 95%, Thermo Scientific Chemicals

Catalog number: 428271000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Phosphorus pentabromide, 95%, Thermo Scientific Chemicals

Catalog number: 428271000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428271000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-69-7
IUPAC Name
pentabromo-λ⁵-phosphane
Molecular Formula
Br5P
InChI Key
QRKVRHZNLKTPGF-UHFFFAOYSA-N
SMILES
BrP(Br)(Br)(Br)Br
Appearance (Color)
Yellow to orange to red
Titration Argentometric
>=94 %
Appearance (Form)
Crystalline powder or crystals and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text