Thermo Scientific Chemicals

Erbium(III) perchlorate, 40 wt.% solution in water, Thermo Scientific Chemicals

Catalog number: 428290250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Erbium(III) perchlorate, 40 wt.% solution in water, Thermo Scientific Chemicals

Catalog number: 428290250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428290250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14017-55-1, 7732-18-5
Titration Complexometric
14 to 15 % (Er)
Appearance (Color)
Clear pink to purple
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text