(±)-Mevalonolactone, 97%, Thermo Scientific Chemicals
(±)-Mevalonolactone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(±)-Mevalonolactone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 428300010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS674-26-0
IUPAC Name4-hydroxy-4-methyloxan-2-one
Molecular FormulaC6H10O3
InChI KeyJYVXNLLUYHCIIH-UHFFFAOYNA-N
SMILESCC1(O)CCOC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
Infrared spectrumConforms
Appearance (Form)Liquid or solid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only