Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 428321000
Price (USD)/ Each
130.00
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
130.00
Add to cart
Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Catalog number428321000
Price (USD)/ Each
130.00
-
Add to cart
Chemical Identifiers
CAS29957-59-3, 75-09-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Solution
Strength0.95 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only