Thermo Scientific Chemicals

Ammonia, ca. 7N solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428381000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Ammonia, ca. 7N solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428381000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 428381000
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7664-41-7, 67-56-1
Strength
6.0 to 7.5 N
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Vapors
    Category 3
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Specific target organ toxicity - (single exposure)
    Category 1
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

    H314 - Causes severe skin burns and eye damage

    H370 - Causes damage to organs

    H412 - Harmful to aquatic life with long lasting effects

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P308 + P311 - IF exposed or concerned: Call a POISON CENTER or doctor

    P310 - Immediately call a POISON CENTER or doctor/physician