Thermo Scientific Chemicals

Methyl cellulose, viscosity 400 cP (2% solution in water), Thermo Scientific Chemicals

Catalog number: 428430500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl cellulose, viscosity 400 cP (2% solution in water), Thermo Scientific Chemicals

Catalog number: 428430500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428430500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-67-5
IUPAC Name
3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular Formula
C20H38O11
InChI Key
YLGXILFCIXHCMC-UHFFFAOYNA-N
SMILES
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
Infrared spectrum
Conforms
Appearance (Color)
White to yellowish-white to greyish-white
Appearance (Form)
Powder
Loss on drying
=<5 %
Viscosity
300 to 560 cP (2 % in water at 20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text