Methyl tricosanoate, 99+%, Thermo Scientific Chemicals
Methyl tricosanoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl tricosanoate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 428450050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2433-97-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
NMR (1H-NMR)Consistent with structure
Infrared spectrumConforms
Melting point53°C to 56°C
Appearance (Form)Powder or solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only