Methyl 1-hydroxy-2-naphthoate, Thermo Scientific Chemicals
Methyl 1-hydroxy-2-naphthoate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 1-hydroxy-2-naphthoate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 428470050
Price (USD)/ Each
141.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
141.00
Add to cart
Methyl 1-hydroxy-2-naphthoate, Thermo Scientific Chemicals
Catalog number428470050
Price (USD)/ Each
141.00
-
Add to cart
Chemical Identifiers
CAS948-03-8
IUPAC Namemethyl 1-hydroxynaphthalene-2-carboxylate
Molecular FormulaC12H10O3
InChI KeyHMIBDRSTVGFJPB-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C2C=CC=CC2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point74°C to 80°C
GC>=97.5 %
Appearance (Color)White to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only