Thermo Scientific Chemicals

Sodium phosphite dibasic pentahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 428502500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium phosphite dibasic pentahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 428502500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428502500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13517-23-2
IUPAC Name
disodium pentahydrate phosphonate
Molecular Formula
H11Na2O8P
InChI Key
VCNBZJAEINSDQB-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Na+].[Na+].[O-]P([O-])=O
Appearance (Color)
Transparent white to white
Titration with HCl
>=97.5 %
Chloride (Cl)
=<50 ppm
Heavy metals (as Pb)
=<10 ppm
Iron (Fe)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text