Glycine benzyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Glycine benzyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycine benzyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass Bottle
Catalog number 428540250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebenzyl 2-aminoacetate hydrochloridyl
Molecular FormulaC9H11ClNO2
InChI KeyBIGFBRPKXPVZGE-UHFFFAOYSA-N
SMILES[Cl].NCC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol)200.64
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystalline powder or solid
Infrared spectrumConforms
NMRConsistent with structure
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only