Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 428561000
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
Add to cart
Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
Catalog number428561000
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7646-85-7, 96-47-9
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Appearance (Color)Clear to hazy colorless to pink to yellow-brown
Strength23 to 27 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only