Thermo Scientific Chemicals

Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium sa, Thermo Scientific Chemicals

Catalog number: 428570500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium sa, Thermo Scientific Chemicals

Catalog number: 428570500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 428570500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13368-13-3
Appearance (Form)
Crystalline powder
Assay
18.46 to 20.82 % (ICP on dried sample)
Infrared spectrum
Conforms
Appearance (Color)
White
Loss on drying
=<20 % (110°C, 16 hrs)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text