1x1 image pixel for data collection
9-Bromo-9-phenylfluorene, 97%
9-Bromo-9-phenylfluorene, 97%
9-Bromo-9-phenylfluorene, 97%
9-Bromo-9-phenylfluorene, 97%
Thermo Scientific Chemicals

9-Bromo-9-phenylfluorene, 97%

CAS: 55135-66-5 | C19H13Br | 321.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4286100505 gGlass bottle
Catalog number 428610050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS55135-66-5
IUPAC Name9-bromo-9-phenyl-9H-fluorene
Molecular FormulaC19H13Br
InChI KeyHYQXNCDBSALQLB-UHFFFAOYSA-N
SMILESBrC1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
HPLC>=96.0 %
Melting point99°C to 103°C
NMR (1H-NMR)Consistent with structure
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link