Chemical Identifiers
CAS21653-40-7
IUPAC Namehydrogen 5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)-2-(phenylformamido)pentanamide chloride
Molecular FormulaC19H23ClN6O4
InChI KeyDEOKFPFLXFNAON-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
View more