Tributyl borate, 98%, AcroSeal™, Thermo Scientific Chemicals
Tributyl borate, 98%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tributyl borate, 98%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 428661000
Price (USD)/ Each
56.60
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
56.60
Add to cart
Tributyl borate, 98%, AcroSeal™, Thermo Scientific Chemicals
Catalog number428661000
Price (USD)/ Each
56.60
-
Add to cart
Chemical Identifiers
CAS688-74-4
IUPAC Nametributyl borate
Molecular FormulaC12H27BO3
InChI KeyLGQXXHMEBUOXRP-UHFFFAOYSA-N
SMILESCCCCOB(OCCCC)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only