Thermo Scientific Chemicals

Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428730010
1 L, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428730010
1 L, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 428730010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-38-6
IUPAC Name
dimethyl carbonate
Molecular Formula
C3H6O3
InChI Key
IEJIGPNLZYLLBP-UHFFFAOYSA-N
SMILES
COC(=O)OC
Color scale
=<30 APHA
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
Water
=<0.005 %
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text