2-Methoxyethanol, for HPLC, Thermo Scientific Chemicals
2-Methoxyethanol, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxyethanol, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number 428780025
Price (USD)/ Each
230.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
271.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
230.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
271.00 
Save 40.35 (15%)
Add to cart
2-Methoxyethanol, for HPLC, Thermo Scientific Chemicals
Catalog number428780025
Price (USD)/ Each
230.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
271.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.05 %
Residue after evaporation=<0.001 %
UV1 cm cell vs HPLC-grade water
UVat 220 nm A: =<1.0
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only