Thermo Scientific Chemicals

Sodium trimethylsilanolate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428841000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Sodium trimethylsilanolate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 428841000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 428841000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18027-10-6
IUPAC Name
sodium trimethylsilanolate
Molecular Formula
C3H9NaOSi
InChI Key
HSNUIYJWTSJUMS-UHFFFAOYSA-N
SMILES
[Na+].C[Si](C)(C)[O-]
Appearance (Color)
Light yellow to amber
Appearance
May develop some turbidity or precipitate
Titration with HClO4
0.90 to 1.10 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text