Potassium hydroxide, 1N solution in ethanol, AcroSeal™, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydroxide, 1N solution in ethanol, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 429091000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3, 64-17-5
SpecificationsSpecification SheetSpecification Sheet
Strength0.950 to 1.050 N
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use