Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 429161000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 429161000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 429161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
368-39-8, 75-09-2
Appearance
May develop some turbidity or precipitate
Appearance (Color)
Clear to slightly hazy colorless to yellow to
Appearance (Form)
Solution
Appearance (Color)
pink to brown
Titration with NaOH
0.90 to 1.10 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkyl transfer reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text