n-Butyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 429230010
Price (USD)/ Each
129.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
152.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
129.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
152.00 
Save 22.35 (15%)
Add to cart
n-Butyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number429230010
Price (USD)/ Each
129.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
152.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.005 %
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only