Anisole, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Anisole, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anisole, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number 429250010
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
220.00 
Save 32.35 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
220.00 
Save 32.35 (15%)
Add to cart
Anisole, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number429250010
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
220.00 
Save 32.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameanisole
Molecular FormulaC7H8O
InChI KeyRDOXTESZEPMUJZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1
Molecular Weight (g/mol)108.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.005 %
Residue after evaporation=<0.005 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only