Thermo Scientific Chemicals

Propylene carbonate, 99.5%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 429268000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Propylene carbonate, 99.5%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 429268000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 429268000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-32-7
IUPAC Name
4-methyl-1,3-dioxolan-2-one
Molecular Formula
C4H6O3
InChI Key
RUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILES
CC1COC(=O)O1
Appearance (Form)
Liquid
GC
>=99.4 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Water
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text