Thermo Scientific Chemicals

Amberlite™ IRN-78 ion-exchange resin, OH-form, Thermo Scientific Chemicals

Catalog number: 429291000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Amberlite™ IRN-78 ion-exchange resin, OH-form, Thermo Scientific Chemicals

Catalog number: 429291000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 429291000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11128-95-3
MDL Number
MFCD00145822
Uniformity coefficient
=<1.20
Total volume capacity
>=1.10 eq/l (OH) as is
Appearance (Form)
Spherical beads
Moisture holding capacity
54.0 to 60.0 % (OH)
Harmonic mean size
0.580 to 0.680 mm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text