1-Methyl-1H-indazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals
1-Methyl-1H-indazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-1H-indazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 429340050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1053655-56-3
IUPAC Name1-methyl-1H-indazole-4-carbaldehyde
Molecular FormulaC9H8N2O
InChI KeySONWSIAHVBTXPB-UHFFFAOYSA-N
SMILESCN1N=CC2=C(C=O)C=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point76°C to 80°C
Appearance (Color)Light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only