1,4-Benzodioxan-5-amine, 95%, Thermo Scientific Chemicals
1,4-Benzodioxan-5-amine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Benzodioxan-5-amine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 429370050
Price (USD)/ Each
610.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
610.00
Add to cart
1,4-Benzodioxan-5-amine, 95%, Thermo Scientific Chemicals
Catalog number429370050
Price (USD)/ Each
610.00
-
Add to cart
Chemical Identifiers
CAS16081-45-1
IUPAC Name2,3-dihydro-1,4-benzodioxin-5-amine
Molecular FormulaC8H9NO2
InChI KeyDMLRSJNZORFCBD-UHFFFAOYSA-N
SMILESNC1=C2OCCOC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear brown to purple
GC>=94.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only