8-Aminoisoquinoline, 97%, Thermo Scientific Chemicals
8-Aminoisoquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Aminoisoquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 429400050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23687-27-6
IUPAC Nameisoquinolin-8-amine
Molecular FormulaC9H8N2
InChI KeyGUSYANXQYUJOBH-UHFFFAOYSA-N
SMILESNC1=C2C=NC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder or solid
Melting point167°C to 170°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only