Thermo Scientific Chemicals

6-Methoxypyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 429410010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Methoxypyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 429410010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 429410010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
312300-42-8
IUPAC Name
6-methoxypyridine-3-sulfonyl chloride
Molecular Formula
C6H6ClNO3S
InChI Key
OMLIVJOTRYWHNY-UHFFFAOYSA-N
SMILES
COC1=NC=C(C=C1)S(Cl)(=O)=O
Appearance (Form)
Low melting crystalline solid
GC
>=96.0 %
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Melting point
42°C to 46°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text