Thermo Scientific Chemicals

Tetrahydropyran-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 429440010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrahydropyran-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 429440010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 429440010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
338453-21-7
IUPAC Name
oxane-4-sulfonyl chloride
Molecular Formula
C5H9ClO3S
InChI Key
QQSBPTQNEOJFBO-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1CCOCC1
Infrared spectrum
Conforms
Appearance (Color)
Colorless to light brown
GC
>=96.0 %
Appearance (Form)
Low melting solid
Melting point
35°C to 41°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text