1H-Benzimidazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
1H-Benzimidazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1H-Benzimidazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 429510050
Price (USD)/ Each
1,079.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
1,270.00 
Save 190.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
1,079.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
1,270.00 
Save 190.35 (15%)
Add to cart
1H-Benzimidazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog number429510050
Price (USD)/ Each
1,079.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
1,270.00 
Save 190.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1H-1,3-benzodiazole-4-carboxylic acid
Molecular FormulaC8H6N2O2
InChI KeyVVQNAFBGAWCMLU-UHFFFAOYSA-N
SMILESOC(=O)C1=C2N=CNC2=CC=C1
Molecular Weight (g/mol)162.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to light brown
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only