Thermo Scientific Chemicals

2-chlorothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 429550050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-chlorothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 429550050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 429550050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5198-87-8
IUPAC Name
2-chloro-1,3-thiazole-4-carboxylic acid
Molecular Formula
C4H2ClNO2S
InChI Key
UVYJJJQMZPCYKY-UHFFFAOYSA-N
SMILES
OC(=O)C1=CSC(Cl)=N1
Appearance (Form)
Powder or crystalline powder
Appearance (Color)
White to yellow to orange
HPLC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text