2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 429570050
Price (USD)/ Each
485.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
485.00
Add to cart
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
Catalog number429570050
Price (USD)/ Each
485.00
-
Add to cart
Chemical Identifiers
CAS40283-46-3
IUPAC Name2-amino-1,3-thiazole-5-carboxylic acid
Molecular FormulaC4H4N2O2S
InChI KeyZFMRDDYYJJCBKC-UHFFFAOYSA-N
SMILESNC1=NC=C(S1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Off-white to yellow
HPLC>=94 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only