Quinoxaline-5-carboxylic acid, 95%, Thermo Scientific Chemicals
Quinoxaline-5-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Quinoxaline-5-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 429580010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6924-66-9
IUPAC Namequinoxaline-5-carboxylic acid
Molecular FormulaC9H6N2O2
InChI KeyQLZNISOPACYKOR-UHFFFAOYSA-N
SMILESOC(=O)C1=C2N=CC=NC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Form)Powder
Melting point166°C to 173°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only