(BOC-aminooxy)acetic acid, 98%, Thermo Scientific Chemicals
(BOC-aminooxy)acetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(BOC-aminooxy)acetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 429690050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS42989-85-5
IUPAC Name2-({[(tert-butoxy)carbonyl]amino}oxy)acetic acid
Molecular FormulaC7H13NO5
InChI KeyQBXODCKYUZNZCY-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NOCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
Melting point110°C to 113°C (Dec)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only